Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101200.2 | NP_001094670.1 | 1 - 2 | 61 | 94 | |
GenBank mRNA | BC151255.1 | - | 1 - 2 | 62 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |