Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101232.1 | NP_001094702.1 | 1 - 2 | 116 | 154 | |
GenBank mRNA | BC149122.1 | - | 1 - 2 | 116 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |