Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB27A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101270.1 | NP_001094740.1 | 2 - 3 | 273 | 76 | |
GenBank mRNA | BC147901.1 | - | 2 - 3 | 273 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |