Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL20RB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102175.2 | NP_001095645.1 | 3 - 4 | 566 | 114 | |
GenBank mRNA | BC148928.1 | - | 3 - 4 | 567 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |