Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102193.1 | NP_001095663.1 | 3 - 4 | 1183 | 58 | |
GenBank mRNA | BT030693.1 | - | 3 - 4 | 1183 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Bos taurus Probe spans exons |