Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYSLTR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102240.2 | NP_001095710.1 | 2 - 3 | 1329 | 122 | |
GenBank mRNA | BC151559.1 | - | 3 - 4 | 1370 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |