Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYTIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102243.2 | NP_001095713.1 | 3 - 4 | 322 | 103 | |
GenBank mRNA | BC151462.1 | - | 3 - 4 | 323 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |