Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYTIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102243.2 | NP_001095713.1 | 4 - 5 | 426 | 74 | |
GenBank mRNA | BC151462.1 | - | 4 - 5 | 427 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |