Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCEANC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104992.2 | NP_001098462.1 | 2 - 3 | 315 | 113 | |
GenBank mRNA | BC142457.1 | - | 2 - 3 | 316 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |