Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DBI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113321.1 | NP_001106792.1 | 2 - 3 | 253 | 67 | |
GenBank mRNA | BC114181.1 | - | 2 - 3 | 228 | 67 | |
M15886.1 | - | 2 - 3 | 253 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |