Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114083.2 | NP_001107555.1 | 1 - 2 | 458 | 59 | |
GenBank mRNA | BC103162.1 | - | 1 - 2 | 459 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |