Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101286.1 | NP_001094756.1 | 2 - 3 | 250 | 127 | |
GenBank mRNA | BC148048.1 | - | 2 - 3 | 250 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |