Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102543.1 | NP_001096013.1 | 5 - 6 | 818 | 74 | |
GenBank mRNA | BC149008.1 | - | 5 - 6 | 818 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |