Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105471.1 | NP_001098941.1 | 1 - 2 | 31 | 56 | |
GenBank mRNA | BC134484.1 | - | 1 - 2 | 31 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |