Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLSCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105506.2 | NP_001098976.2 | 1 - 2 | 88 | 135 | |
GenBank mRNA | BC149269.1 | - | 1 - 2 | 89 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |