Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075382.2 | NP_001068850.2 | 2 - 3 | 442 | 109 | |
NM_001105456.2 | NP_001098926.1 | 2 - 3 | 442 | 109 | ||
GenBank mRNA | BC149743.1 | - | 2 - 3 | 443 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |