Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAG17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176627.1 | NP_788800.1 | 2 - 3 | 208 | 151 | |
GenBank mRNA | AB098938.1 | - | 2 - 3 | 246 | 151 | |
AB098948.1 | - | 2 - 3 | 208 | 150 | ||
AB099090.1 | - | 1 - 2 | 93 | 151 | ||
AF192334.1 | - | 2 - 3 | 208 | 151 | ||
BC134714.1 | - | 2 - 3 | 257 | 151 | ||
BC151448.1 | - | 2 - 3 | 257 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |