Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REPIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075521.1 | NP_001068989.1 | 4 | 2347 | 133 | |
GenBank mRNA | BC120236.1 | - | 4 | 2347 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |