Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031756.2 | NP_001026926.1 | 1 - 2 | 235 | 93 | |
GenBank mRNA | AY911360.1 | - | 2 - 3 | 253 | 93 | |
BC109856.1 | - | 1 | 253 | 93 | ||
BC133430.1 | - | 2 - 3 | 277 | 93 | ||
BT021543.1 | - | 1 - 2 | 238 | 93 | ||
BT021544.1 | - | 1 - 2 | 238 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |