Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HACD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130757.2 | NP_001124229.1 | 6 - 7 | 757 | 128 | |
GenBank mRNA | BC112557.1 | - | 6 - 7 | 759 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |