Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130760.1 | NP_001124232.1 | 3 - 4 | 297 | 53 | |
GenBank mRNA | BC108116.1 | - | 3 - 4 | 297 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |