Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099170.1 | NP_001092640.1 | 6 - 7 | 714 | 89 | |
GenBank mRNA | BC146095.1 | - | 6 - 7 | 714 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |