Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TWF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105324.2 | NP_001098794.1 | 8 - 9 | 1019 | 98 | |
GenBank mRNA | AY124008.1 | - | 3 - 4 | 305 | 98 | |
BC133441.1 | - | 8 - 9 | 1021 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |