Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDYL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163785.1 | NP_001157257.1 | 2 - 3 | 638 | 114 | |
GenBank mRNA | EF690283.1 | - | 2 - 3 | 638 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |