Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205425.3 | NP_001192354.3 | 1 - 2 | 34 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |