Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JPH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205619.1 | NP_001192548.1 | 4 - 5 | 2183 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Bos taurus Probe spans exons |