Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205698.1 | NP_001192627.1 | 11 - 12 | 2548 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |