Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHPF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205751.2 | NP_001192680.2 | 1 - 2 | 606 | 105 | |
GenBank mRNA | BC148920.1 | - | 1 - 2 | 338 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |