Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205962.2 | NP_001192891.2 | 4 - 5 | 571 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Bos taurus Probe spans exons |