Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205993.2 | NP_001192922.1 | 5 - 6 | 898 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Bos taurus Probe spans exons |