Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206280.3 | NP_001193209.2 | 2 - 3 | 332 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |