Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206378.1 | NP_001193307.1 | 5 - 6 | 949 | 75 | |
GenBank mRNA | BC134436.1 | - | 5 - 6 | 950 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |