Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIVM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206453.2 | NP_001193382.2 | 7 - 8 | 1538 | 90 | |
GenBank mRNA | EE907980.1 | - | 5 - 6 | 541 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |