Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AASDHPPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206477.1 | NP_001193406.1 | 1 - 2 | 257 | 116 | |
GenBank mRNA | BC102625.1 | - | 1 - 2 | 226 | 116 | |
DT813625.1 | - | 1 - 2 | 179 | 116 | ||
DT860683.1 | - | 1 - 2 | 258 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |