Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MANEA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206725.1 | NP_001193654.1 | 3 - 4 | 797 | 111 | |
GenBank mRNA | BC103138.1 | - | 3 - 4 | 799 | 111 | |
JN385276.1 | - | 2 - 3 | 681 | 111 | ||
JQ946519.1 | - | 2 - 3 | 654 | 111 | ||
JQ946520.1 | - | 2 - 3 | 654 | 111 | ||
JQ946521.1 | - | 2 - 3 | 654 | 111 | ||
JQ946522.1 | - | 2 - 3 | 654 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |