Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERO1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206730.1 | NP_001193659.1 | 11 - 12 | 809 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |