Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206778.1 | NP_001193707.1 | 8 - 9 | 1201 | 57 | |
NM_001206779.1 | NP_001193708.1 | 6 - 7 | 763 | 57 | ||
GenBank mRNA | CK964625.1 | - | 2 - 3 | 102 | 57 | |
CK981807.1 | - | 3 - 4 | 213 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Bos taurus Probe spans exons |