Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTOP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206784.2 | NP_001193713.1 | 2 - 3 | 551 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |