Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTFMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166523.3 | NP_001159995.1 | 8 - 9 | 992 | 101 | |
GenBank mRNA | AB004316.1 | - | 8 - 9 | 927 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Bos taurus Probe spans exons |