Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GYPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001184951.1 | NP_001171880.1 | 2 - 3 | 167 | 63 | |
GenBank mRNA | AB558590.2 | - | 2 - 3 | 167 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |