Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPDYA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191147.2 | NP_001178076.2 | 5 - 6 | 652 | 111 | |
GenBank mRNA | BC109600.1 | - | 5 - 6 | 636 | 111 | |
DT833851.1 | - | 5 - 6 | 654 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |