Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191173.2 | NP_001178102.1 | 6 - 7 | 434 | 73 | |
GenBank mRNA | BC142461.1 | - | 6 - 7 | 409 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |