Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191268.1 | NP_001178197.1 | 6 - 7 | 3350 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |