Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242955.2 | NP_001229884.1 | 1 - 2 | 88 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |