Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDGFL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191368.2 | NP_001178297.1 | 5 - 6 | 945 | 92 | |
GenBank mRNA | EH202588.1 | - | 4 - 5 | 480 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Bos taurus Probe spans exons |