Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PABPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174568.3 | NP_776993.1 | 11 - 12 | 2115 | 78 | |
GenBank mRNA | AJ401269.2 | - | 11 - 12 | 1601 | 78 | |
BC102365.1 | - | 11 - 12 | 2105 | 78 | ||
EE252871.1 | - | 4 - 5 | 600 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |