Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PABPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174568.3 | NP_776993.1 | 14 - 15 | 2429 | 85 | |
GenBank mRNA | BC102365.1 | - | 14 - 15 | 2419 | 85 | |
DV850814.1 | - | 3 - 4 | 249 | 85 |
Gene Symbol: PABPC1P10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | U83076.1 | - | 1 - 2 | 204 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |