Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLVCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192143.1 | NP_001179072.1 | 6 - 7 | 1128 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |