Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192291.2 | NP_001179220.1 | 2 - 3 | 753 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |