Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GXYLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193167.1 | NP_001180096.1 | 5 - 6 | 868 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |